Assay Details
| Gene Symbol: Smpd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031360.2 | NP_112650.1 | 1 - 2 | 310 | 63 | |
| XM_008773007.1 | XP_008771229.1 | 2 - 3 | 247 | 63 | ||
| GenBank mRNA | AB047002.1 | - | 1 - 2 | 49 | 63 | |
| BC091139.1 | - | 1 - 2 | 310 | 7378100 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smpd2
Genomic Map