Assay Details
| Gene Symbol: Ccl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031530.1 | NP_113718.1 | 1 - 2 | 155 | 95 | |
| GenBank mRNA | AF058786.1 | - | 1 - 2 | 155 | 95 | |
| M57441.1 | - | 1 - 2 | 119 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Citations (3)
SOCS3-mediated blockade of JAK/STAT3 signaling pathway reveals its major contribution to spinal cord neuroinflammation and mechanical allodynia after peripheral nerve injury.
- Authors
- Dominguez E, Mauborgne A, Mallet J, Desclaux M, Pohl M
- Journal
- J Neurosci - Page No 5754 (Wed Apr 21 00:00:00 UTC 2010)
- Volume (Year)
- 30 (2010)
- Product Usage
- -
Effects of a new bioactive lipid-based drug carrier on cultured hepatic stellate cells and liver fibrosis in bile duct-ligated rats.
- Authors
- Adrian JE, Poelstra K, Scherphof GL, Meijer DK, van Loenen-Weemaes AM, Reker-Smit C, Morselt HW, Zwiers P, Kamps JA
- Journal
- J Pharmacol Exp Ther - Page No 536 (Tue May 01 00:00:00 UTC 2007)
- Volume (Year)
- 321 (2007)
- Product Usage
- -
Early postnatal oestradiol exposure causes insulin resistance and signs of inflammation in circulation and skeletal muscle.
- Authors
- Alexanderson C, Eriksson E, Stener-Victorin E, Lonn M, Holmang A
- Journal
- J Endocrinol - Page No 49 (Wed Apr 01 00:00:00 UTC 2009)
- Volume (Year)
- 201 (2009)
- Product Usage
- -

Ccl2
Genomic Map