Assay Details
| Gene Symbol: Eif2ak3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031599.2 | NP_113787.1 | 12 - 13 | 2226 | 148 | |
| XM_006236623.1 | XP_006236685.1 | 12 - 13 | 1744 | 148 | ||
| XM_006236624.2 | XP_006236686.1 | 12 - 13 | 2264 | 148 | ||
| XM_008762977.1 | XP_008761199.1 | 11 - 12 | 1858 | 148 | ||
| GenBank mRNA | AF096835.1 | - | 12 - 13 | 2227 | 148 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
ossification
angiogenesis
chondrocyte development
translation
protein phosphorylation
activation of cysteine-type endopeptidase activity involved in apoptotic process
ER overload response
endoplasmic reticulum organization
lactation
positive regulation of signal transduction
positive regulation of vascular endothelial growth factor production
positive regulation of gene expression
negative regulation of gene expression
regulation of translational initiation by eIF2 alpha phosphorylation
negative regulation of translation
regulation of fatty acid metabolic process
calcium-mediated signaling
bone mineralization
endoplasmic reticulum unfolded protein response
pancreas development
endocrine pancreas development
negative regulation of myelination
negative regulation of translation in response to stress
negative regulation of translational initiation in response to stress
positive regulation of protein binding
SREBP signaling pathway
response to endoplasmic reticulum stress
cellular response to glucose starvation
negative regulation of apoptotic process
fat cell differentiation
positive regulation of transcription from RNA polymerase I promoter
protein autophosphorylation
insulin-like growth factor receptor signaling pathway
protein homooligomerization
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
positive regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
protein kinase activity
protein serine/threonine kinase activity
eukaryotic translation initiation factor 2alpha kinase activity
ATP binding
enzyme binding
protein phosphatase binding
identical protein binding
skeletal system development
ossification
angiogenesis
chondrocyte development
translation
protein phosphorylation
activation of cysteine-type endopeptidase activity involved in apoptotic process
ER overload response
endoplasmic reticulum organization
lactation
positive regulation of signal transduction
positive regulation of vascular endothelial growth factor production
positive regulation of gene expression
negative regulation of gene expression
regulation of translational initiation by eIF2 alpha phosphorylation
negative regulation of translation
regulation of fatty acid metabolic process
calcium-mediated signaling
bone mineralization
endoplasmic reticulum unfolded protein response
pancreas development
endocrine pancreas development
negative regulation of myelination
negative regulation of translation in response to stress
negative regulation of translational initiation in response to stress
positive regulation of protein binding
SREBP signaling pathway
response to endoplasmic reticulum stress
cellular response to glucose starvation
negative regulation of apoptotic process
fat cell differentiation
positive regulation of transcription from RNA polymerase I promoter
protein autophosphorylation
insulin-like growth factor receptor signaling pathway
protein homooligomerization
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
positive regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
protein kinase activity
protein serine/threonine kinase activity
eukaryotic translation initiation factor 2alpha kinase activity
ATP binding
enzyme binding
protein phosphatase binding
identical protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P629500 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759085 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147588 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616496 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949457 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965734 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152349 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946214 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41245 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806572 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407094 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24590 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297243 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414689 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110509 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715628 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39066 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163069 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406146 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60803 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491448 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220329 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379165 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407387 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613667 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154477 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36717 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510903 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181267 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570900 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909095 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573082 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9981 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835838 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355264 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785446 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342180 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665401 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312228 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15183 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148166 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70291 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99856 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8468 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11951 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173819 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P629500 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P629500 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P629500 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P466200 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701202235 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 5901 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 22450 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 66344 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 70505 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 16045 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 52642 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 701202235 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10856248 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10856243 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10856247 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10856242 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10856245 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10856236 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10856249 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10856234 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10856233 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10856246 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10856240 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10856241 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10856235 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10856237 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10856239 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10856238 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10856244 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10856232 | - | - |
| Rat Genome 230 2.0 Array | 1368326_at | - | - |
| Rat Genome 230 2.0 Array | 1368326_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368326_at | - | - |
| Affymetrix HT Rat Focus Array | 1368326_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0400007695.rn.1 | TC0400001093.rn.1 | RTA TC |
| Rat Expression Set 230 | 1368326_at | - | - |
| Rat Expression Set 230 | 1368326_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10856232 | - | - |
| Rat Genome U34 Set | AF096835_g_at | - | - |
| Rat Genome U34 Set | AF096835_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20840924 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1374121 | - | - |
| RatRef-12 Expression BeadChip | ILMN_63795 | - | - |

Eif2ak3
Genomic Map