Assay Details
| Gene Symbol: Ramp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031645.1 | NP_113833.1 | 1 - 2 | 93 | 115 | |
| GenBank mRNA | AB042887.1 | - | 1 - 2 | 93 | 115 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB042887.1, NM_031645.1 transcripts.
Set Membership: |
|

Ramp1
Genomic Map