Assay Details
| Gene Symbol: Stk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031735.1 | NP_113923.1 | 8 - 9 | 1032 | 86 | |
| XM_008765458.1 | XP_008763680.1 | 8 - 9 | 1210 | 86 | ||
| XM_008765459.1 | XP_008763681.1 | 8 - 9 | 1210 | 86 | ||
| XM_008765460.1 | XP_008763682.1 | 8 - 9 | 899 | 86 | ||
| XM_008765461.1 | XP_008763683.1 | 6 - 7 | 877 | 86 | ||
| XM_008765462.1 | XP_008763684.1 | 6 - 7 | 877 | 86 | ||
| GenBank mRNA | AJ001529.1 | - | 8 - 9 | 1032 | 86 | |
| BC161844.1 | - | 8 - 9 | 1115 | 9027884 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
neural tube formation
endocardium development
protein phosphorylation
apoptotic process
central nervous system development
negative regulation of cell proliferation
signal transduction by protein phosphorylation
positive regulation of protein binding
hippo signaling
intracellular signal transduction
positive regulation of apoptotic process
positive regulation of fat cell differentiation
positive regulation of JNK cascade
negative regulation of organ growth
protein stabilization
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of protein kinase B signaling
primitive hemopoiesis
cell differentiation involved in embryonic placenta development
regulation of cell differentiation involved in embryonic placenta development
negative regulation of canonical Wnt signaling pathway
hepatocyte apoptotic process
positive regulation of extrinsic apoptotic signaling pathway via death domain receptors
magnesium ion binding
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
ATP binding
metal ion binding
neural tube formation
endocardium development
protein phosphorylation
apoptotic process
central nervous system development
negative regulation of cell proliferation
signal transduction by protein phosphorylation
positive regulation of protein binding
hippo signaling
intracellular signal transduction
positive regulation of apoptotic process
positive regulation of fat cell differentiation
positive regulation of JNK cascade
negative regulation of organ growth
protein stabilization
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of protein kinase B signaling
primitive hemopoiesis
cell differentiation involved in embryonic placenta development
regulation of cell differentiation involved in embryonic placenta development
negative regulation of canonical Wnt signaling pathway
hepatocyte apoptotic process
positive regulation of extrinsic apoptotic signaling pathway via death domain receptors
magnesium ion binding
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
ATP binding
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P525024 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P164716 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20978 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479048 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721789 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408815 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211264 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498417 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766182 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314681 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31047 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442580 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210398 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326987 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950082 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815651 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412472 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712260 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800066 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353994 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310595 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235200 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971493 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109151 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929758 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588773 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90096 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837952 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240767 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585240 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313609 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384039 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654093 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963261 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303940 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347807 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228628 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907150 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420467 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953019 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794051 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805242 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137781 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871599 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74922 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249361 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114731 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216065 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188348 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812522 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262324 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593639 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170788 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397132 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45365 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313642 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142486 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597307 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171171 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909584 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50350 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740825 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891361 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926987 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269890 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189437 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913944 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383353 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776385 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282600 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74103 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244817 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933260 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372475 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699186 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 81420 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839310 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337109 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192107 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215768 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365991 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11382 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181321 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198005 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947851 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242904 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256897 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228384 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116248 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129744 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726900 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536808 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939079 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696809 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687454 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720470 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854423 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814879 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181820 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825970 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431114 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152069 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542149 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646524 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288859 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393148 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694380 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12957 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403748 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485545 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296225 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698620 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853674 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562348 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407004 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288378 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222382 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102480 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239052 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147207 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 713430 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834667 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343110 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517838 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214991 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10003 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691545 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558812 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106858 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731027 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94859 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758343 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225331 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618992 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843862 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930729 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754199 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647960 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649195 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780625 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303402 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397537 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171366 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162117 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53653 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 45919 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5211 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9777 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96069 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 29184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35509 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133761 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70036 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120251 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78436 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101351 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20279 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 103244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139215 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109163 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121435 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175598 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141432 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168714 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155230 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153200 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 177681 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P164716 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P525024 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P525024 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P164716 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P525024 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P164716 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P525024 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700362550 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 82387 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 104217 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 16185 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 89740 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 66420 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 99128 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 51942 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 39639 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 56167 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 60027 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 5549 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 32506 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 63449 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 82545 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 61828 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 55275 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 80615 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 20799 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 9196 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 94668 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700362550 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10903257 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10903248 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10903250 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10903258 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10903255 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10903249 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10903252 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10903254 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10903247 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10903253 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10903256 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10903251 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10903246 | - | - |
| Rat Genome 230 2.0 Array | 1369712_at | - | - |
| Rat Genome 230 2.0 Array | 1369712_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1369712_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1369712_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0700022975.rn.1 | TC0700002967.rn.1 | RTA TC |
| Rat Expression Set 230 | 1369712_at | - | - |
| Rat Expression Set 230 | 1369712_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10903246 | - | - |
| Rat Genome U34 Set | AJ001529cds_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20908482 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373576 | - | - |
| RatRef-12 Expression BeadChip | ILMN_53175 | - | - |

Stk3
Genomic Map