Assay Details
| Gene Symbol: Nfe2l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031789.2 | NP_113977.1 | 2 - 3 | 379 | 104 | |
| XM_006234396.2 | XP_006234458.1 | 2 - 3 | 527 | 104 | ||
| XM_006234397.2 | XP_006234459.1 | 2 - 3 | 472 | 104 | ||
| XM_006234398.2 | XP_006234460.1 | 2 - 3 | 1079 | 104 | ||
| GenBank mRNA | AF037350.1 | - | 2 - 3 | 381 | 104 | |
| BC061724.1 | - | 2 - 3 | 394 | 5598313 | 104 | |
| FQ221441.1 | - | 2 - 3 | 400 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nfe2l2
Genomic Map