Assay Details
| Gene Symbol: Hspa1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031971.2 | NP_114177.2 | 3023 | 88 | ||
| GenBank mRNA | L16764.1 | - | 1953 | 88 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hspa1a
Genomic Map