Assay Details
| Gene Symbol: Msra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053307.2 | NP_445759.1 | 1 - 2 | 145 | 59 | |
| XM_008770745.1 | XP_008768967.1 | 1 - 2 | 516 | 59 | ||
| GenBank mRNA | AY005464.1 | - | 1 - 2 | 135 | 59 | |
| BC087009.1 | - | 1 - 2 | 145 | 7107102 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P12758 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401921 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258202 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409644 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296565 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618958 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142845 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460193 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351125 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377007 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637883 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50200 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949620 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454375 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953618 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762429 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851391 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498603 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685657 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246978 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221242 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249253 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387435 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209361 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874117 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120402 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377745 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150241 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835895 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714868 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946855 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886181 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480005 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8836 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938707 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50165 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139061 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779262 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387077 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498980 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592374 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511158 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210707 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349081 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420538 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886636 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312020 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801719 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87202 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699618 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85653 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542362 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127328 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403108 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892054 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296493 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854273 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875191 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238183 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520036 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101041 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728117 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185791 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230350 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740780 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465913 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564714 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383260 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 658063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459742 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535097 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93381 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905379 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438904 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501381 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962479 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790317 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291095 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29583 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14633 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362293 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583095 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129122 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793820 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540052 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793577 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756092 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165808 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319583 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233388 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912621 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806001 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 155067 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161243 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145326 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447741 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481565 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792956 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967950 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926199 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112463 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21141 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700397 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973539 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702970 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46743 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946025 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207001 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145345 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276422 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126307 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271206 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272853 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206072 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237663 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767349 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253654 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478102 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752731 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236594 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472992 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274012 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729833 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608012 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848451 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683741 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240170 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464430 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138508 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691150 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838569 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227553 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764898 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854338 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961647 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809331 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10500 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120004 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129586 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176768 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69465 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122068 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62913 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157180 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79903 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101432 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143987 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50791 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24706 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20752 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111843 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140387 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160050 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167983 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P010902 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P010902 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P010902 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P12758 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 57423 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 35168 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 23305 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 73709 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 79449 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 1685 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 29047 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 31408 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 71295 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 58136 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 48972 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 92219 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 81495 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 41151 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 85725 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 6329 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10784522 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784521 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784518 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784519 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784520 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10784517 | - | - |
| Rat Genome 230 2.0 Array | 1387531_at | - | - |
| Rat Genome 230 2.0 Array | 1387531_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387531_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387531_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1500013061.rn.1 | TC1500001921.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1500013052.rn.1 | TC1500001921.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1500013070.rn.1 | TC1500001921.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1500013063.rn.1 | TC1500001921.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1500013060.rn.1 | TC1500001921.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1500013062.rn.1 | TC1500001921.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1500013064.rn.1 | TC1500001921.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1500013069.rn.1 | TC1500001921.rn.1 | RTA TC |
| Rat Expression Set 230 | 1387531_at | - | - |
| Rat Expression Set 230 | 1387531_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784517 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21486630 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1650581 | - | - |

Msra
Genomic Map