Assay Details
| Gene Symbol: Ackr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053352.1 | NP_445804.1 | 1 - 2 | 66 | 67 | |
| GenBank mRNA | AF118816.1 | - | 1 - 2 | 66 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF118816.1, NM_053352.1 transcripts.
Set Membership: |
|

Ackr3
Genomic Map