Assay Details
| Gene Symbol: Slc27a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053580.2 | NP_446032.2 | 7 - 8 | 1173 | 63 | |
| XM_006252953.2 | XP_006253015.1 | 7 - 8 | 1211 | 63 | ||
| XM_006252954.1 | XP_006253016.1 | 7 - 8 | 1159 | 63 | ||
| XM_008771156.1 | XP_008769378.1 | 7 - 8 | 1098 | 63 | ||
| XM_008771157.1 | XP_008769379.1 | 7 - 8 | 1257 | 63 | ||
| GenBank mRNA | BC074014.1 | - | 7 - 8 | 1173 | 7104661 | 63 |
| U89529.1 | - | 7 - 8 | 1070 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc27a1
Genomic Map