Assay Details
| Gene Symbol: F2rl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053808.1 | NP_446260.1 | 1 - 2 | 80 | 111 | |
| GenBank mRNA | AF269246.2 | - | 1 - 2 | 80 | 111 | |
| AF310216.1 | - | 1 - 2 | 107 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

F2rl3
Genomic Map