Assay Details
| Gene Symbol: Cdc123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053877.3 | NP_446329.1 | 4 - 5 | 281 | 90 | |
| GenBank mRNA | BC061527.1 | - | 4 - 5 | 282 | 6920755 | 90 |
| U34843.1 | - | 4 - 5 | 293 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P1051965 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901179 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135999 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733554 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892549 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29870 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491016 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200640 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559867 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160259 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565265 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363618 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624703 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346289 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433238 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760938 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649151 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913950 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828605 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874455 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820209 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973263 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587294 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858384 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46363 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379558 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67125 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96942 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58754 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174306 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1051965 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1051965 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P1051965 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1051965 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700765432 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700484749 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 19142 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 6180 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 66997 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 99203 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700765432 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700484749 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10796299 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796305 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796294 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796306 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796303 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796293 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796297 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796300 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796304 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796295 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796298 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796296 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796301 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796302 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10796292 | - | - |
| Rat Genome 230 2.0 Array | 1381889_at | - | - |
| Rat Genome 230 2.0 Array | 1388105_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1381889_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1388105_at | - | - |
| Affymetrix HT Rat Focus Array | 1381889_at | - | - |
| Affymetrix HT Rat Focus Array | 1388105_at | - | - |
| Affymetrix HT Rat Focus Array | 1388105_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1381889_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1700006806.rn.1 | TC1700000989.rn.1 | RTA TC |
| Rat Expression Set 230 | 1388105_PM_at | - | - |
| Rat Expression Set 230 | 1388105_at | - | - |
| Rat Expression Set 230 | 1381889_at | - | - |
| Rat Expression Set 230 | 1381889_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796292 | - | - |
| Rat Genome U34 Set | U34843_at | - | - |
| Rat Genome U34 Set | U34843_g_at | - | - |
| Rat Genome U34 Set | rc_AA849533_at | - | - |
| Rat Genome U34 Set | rc_AI229968_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21150398 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1351059 | - | - |
| RatRef-12 Expression BeadChip | ILMN_59956 | - | - |

Cdc123
Genomic Map