Assay Details
| Gene Symbol: Tmem158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057212.1 | NP_476560.1 | 1 | 151 | 128 | |
| GenBank mRNA | AB028891.1 | - | 1 | 151 | 128 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB028891.1, NM_057212.1 transcripts.
Set Membership: |
|

Tmem158
Genomic Map