Assay Details
| Gene Symbol: Mgea5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131904.1 | NP_571979.1 | 12 - 13 | 2175 | 111 | |
| GenBank mRNA | AY039679.1 | - | 12 - 13 | 2175 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
N-acetylglucosamine metabolic process
protein deglycosylation
protein targeting to membrane
aging
positive regulation of calcium ion transport into cytosol
negative regulation of cardiac muscle adaptation
histone acetylation
positive regulation of cell killing
positive regulation of insulin secretion
positive regulation of protein complex disassembly
positive regulation of proteolysis
dATP metabolic process
positive regulation of glucose import
response to steroid hormone
positive regulation of DNA metabolic process
positive regulation of mitochondrial depolarization
positive regulation of calcium ion transport
negative regulation of protein glycosylation
positive regulation of growth hormone secretion
necrotic cell death
histone acetyltransferase activity
beta-N-acetylglucosaminidase activity
N-acetylglucosamine metabolic process
protein deglycosylation
protein targeting to membrane
aging
positive regulation of calcium ion transport into cytosol
negative regulation of cardiac muscle adaptation
histone acetylation
positive regulation of cell killing
positive regulation of insulin secretion
positive regulation of protein complex disassembly
positive regulation of proteolysis
dATP metabolic process
positive regulation of glucose import
response to steroid hormone
positive regulation of DNA metabolic process
positive regulation of mitochondrial depolarization
positive regulation of calcium ion transport
negative regulation of protein glycosylation
positive regulation of growth hormone secretion
necrotic cell death
histone acetyltransferase activity
beta-N-acetylglucosaminidase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P15356 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293507 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289496 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273493 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732935 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164322 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4285 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222227 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741778 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831552 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808181 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788698 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121740 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631480 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35435 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566197 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362995 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393297 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106856 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775054 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202717 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154578 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43973 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96193 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161051 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P15356 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P15356 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P15356 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P15356 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 21398 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 1392 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 46052 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 13 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10730467 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730456 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730462 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730468 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730455 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730469 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730466 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730461 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730458 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730463 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730454 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730464 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730459 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730453 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730465 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10730452 | - | - |
| Rat Genome 230 2.0 Array | 1370129_at | - | - |
| Rat Genome 230 2.0 Array | 1398969_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1398969_at | - | - |
| Rat Genome 230 2.0 Array | 1383025_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1370129_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1395741_at | - | - |
| Rat Genome 230 2.0 Array | 1395741_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1383025_at | - | - |
| Affymetrix HT Rat Focus Array | 1383025_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1383025_at | - | - |
| Affymetrix HT Rat Focus Array | 1370129_at | - | - |
| Affymetrix HT Rat Focus Array | 1395741_at | - | - |
| Affymetrix HT Rat Focus Array | 1370129_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1395741_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100062882.rn.1 | TC0100007800.rn.1 | RTA TC |
| Rat Expression Set 230 | 1370129_at | - | - |
| Rat Expression Set 230 | 1370129_PM_at | - | - |
| Rat Expression Set 230 | 1395741_at | - | - |
| Rat Expression Set 230 | 1383025_PM_at | - | - |
| Rat Expression Set 230 | 1395741_PM_at | - | - |
| Rat Expression Set 230 | 1383025_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730452 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21383621 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1370257 | - | - |
| RatRef-12 Expression BeadChip | ILMN_62969 | - | - |

Mgea5
Genomic Map