Assay Details
| Gene Symbol: Ppp1r14c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133425.3 | NP_596916.1 | 3 - 4 | 1022 | 85 | |
| GenBank mRNA | AF407168.3 | - | 3 - 4 | 1022 | 85 | |
| BC086978.1 | - | 3 - 4 | 495 | 7191557 | 85 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P159063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479853 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379050 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93737 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594477 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631470 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510577 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812521 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240859 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187607 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524001 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862818 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438798 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92384 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281475 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75539 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364796 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11343 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702018 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29819 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406345 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122965 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470287 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857282 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957766 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911868 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642327 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954229 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647265 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790305 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912300 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462740 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190191 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781928 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585780 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739716 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622448 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319439 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889340 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649150 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881195 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273099 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872314 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220294 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944901 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141478 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229049 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25226 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396363 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18422 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467041 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731826 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811028 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137495 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370435 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848828 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774683 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78694 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59655 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115827 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140001 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80795 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73354 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53663 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143864 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71296 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136595 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152966 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144193 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71057 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P159063 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P159063 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P159063 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P13189 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P158599 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 80993 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 65713 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 82737 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 9771 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 92249 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 96511 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 5273 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10702600 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702599 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702602 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702604 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702603 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702601 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10702598 | - | - |
| Rat Genome 230 2.0 Array | 1393082_at | - | - |
| Rat Genome 230 2.0 Array | 1368716_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1368716_at | - | - |
| Rat Genome 230 2.0 Array | 1393082_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1393082_at | - | - |
| Affymetrix HT Rat Focus Array | 1368716_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368716_at | - | - |
| Affymetrix HT Rat Focus Array | 1393082_PM_at | - | - |
| Rat Expression Set 230 | 1368716_PM_at | - | - |
| Rat Expression Set 230 | 1368716_at | - | - |
| Rat Expression Set 230 | 1393082_at | - | - |
| Rat Expression Set 230 | 1393082_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702598 | - | - |
| Rat Genome U34 Set | rc_AI044747_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21167150 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1374470 | - | - |
| RatRef-12 Expression BeadChip | ILMN_65990 | - | - |

Ppp1r14c
Genomic Map