Assay Details
| Gene Symbol: Pla2g4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133551.2 | NP_598235.2 | 10 - 11 | 1200 | 84 | |
| GenBank mRNA | BC070940.1 | - | 10 - 11 | 1200 | 7099813 | 84 |
| S77829.1 | - | 10 - 11 | 1186 | 84 | ||
| U38376.1 | - | 10 - 11 | 1201 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
ovulation from ovarian follicle
luteolysis
aging
positive regulation of cell proliferation
phospholipid catabolic process
response to heat
response to hormone
response to organic substance
response to lithium ion
response to organonitrogen compound
arachidonic acid metabolic process
positive regulation of bone mineralization
positive regulation of vesicle fusion
positive regulation of prostaglandin biosynthetic process
positive regulation of fever generation
response to lipopolysaccharide
response to vitamin D
regulation of cell proliferation
response to hydrogen peroxide
positive regulation of apoptotic process
surfactant homeostasis
icosanoid biosynthetic process
decidualization
arachidonic acid secretion
positive regulation of inflammatory response
response to glucocorticoid
response to calcium ion
response to methylmercury
cellular response to antibiotic
lysophospholipase activity
phospholipase A2 activity
calcium ion binding
calcium-dependent phospholipid binding
histone acetyltransferase binding
calcium-dependent phospholipase A2 activity
ovulation from ovarian follicle
luteolysis
aging
positive regulation of cell proliferation
phospholipid catabolic process
response to heat
response to hormone
response to organic substance
response to lithium ion
response to organonitrogen compound
arachidonic acid metabolic process
positive regulation of bone mineralization
positive regulation of vesicle fusion
positive regulation of prostaglandin biosynthetic process
positive regulation of fever generation
response to lipopolysaccharide
response to vitamin D
regulation of cell proliferation
response to hydrogen peroxide
positive regulation of apoptotic process
surfactant homeostasis
icosanoid biosynthetic process
decidualization
arachidonic acid secretion
positive regulation of inflammatory response
response to glucocorticoid
response to calcium ion
response to methylmercury
cellular response to antibiotic
lysophospholipase activity
phospholipase A2 activity
calcium ion binding
calcium-dependent phospholipid binding
histone acetyltransferase binding
calcium-dependent phospholipase A2 activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P477620 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462792 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973847 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747364 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521605 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64252 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854600 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127633 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440689 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488693 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388215 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537354 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812939 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592492 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73560 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290726 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572905 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792824 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635145 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423822 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152289 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477457 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669654 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676811 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858704 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569220 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201769 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912286 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215550 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67132 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303671 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872114 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363624 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961228 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117348 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547169 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912514 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783710 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38850 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567856 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274795 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799293 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880156 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94921 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859402 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186317 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292209 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134645 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135856 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449514 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16468 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552565 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475683 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608143 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701257 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199258 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389578 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508745 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544852 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546623 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482317 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789629 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160068 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523960 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958391 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497167 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440550 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715373 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216889 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937204 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119215 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948432 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243749 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165642 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223398 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431488 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39511 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277566 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959050 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863013 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763937 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303759 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880483 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27687 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388481 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497182 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567952 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923682 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656966 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23371 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19300 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120097 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71489 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40429 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63010 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106340 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65450 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113540 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169068 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174181 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86440 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33861 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160480 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17879 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P477620 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P477620 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P477620 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P477620 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 100346 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 73261 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 5539 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 91939 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 62112 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 82971 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 65835 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 38037 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 73216 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 103007 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 96067 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 1675 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 26162 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10768378 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768386 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768377 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768379 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768382 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768392 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768383 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768393 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768385 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768381 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768387 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768390 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768389 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768394 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768388 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768391 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768384 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768380 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10768376 | - | - |
| Rat Genome 230 2.0 Array | 1387566_at | - | - |
| Rat Genome 230 2.0 Array | 1387566_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387566_at | - | - |
| Affymetrix HT Rat Focus Array | 1387566_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1300010832.rn.1 | TC1300001598.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300010822.rn.1 | TC1300001598.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300010824.rn.1 | TC1300001598.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300010831.rn.1 | TC1300001598.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300010817.rn.1 | TC1300001598.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300010826.rn.1 | TC1300001598.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300010830.rn.1 | TC1300001598.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300010827.rn.1 | TC1300001598.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300010811.rn.1 | TC1300001598.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300010815.rn.1 | TC1300001598.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300010825.rn.1 | TC1300001598.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300010821.rn.1 | TC1300001598.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300010823.rn.1 | TC1300001598.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300010812.rn.1 | TC1300001598.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300010828.rn.1 | TC1300001598.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300010816.rn.1 | TC1300001598.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300010819.rn.1 | TC1300001598.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300010814.rn.1 | TC1300001598.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300010820.rn.1 | TC1300001598.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300010818.rn.1 | TC1300001598.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300010813.rn.1 | TC1300001598.rn.1 | RTA TC |
| Rat Expression Set 230 | 1387566_at | - | - |
| Rat Expression Set 230 | 1387566_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768376 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21565934 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1360060 | - | - |
| RatRef-12 Expression BeadChip | ILMN_60311 | - | - |

Pla2g4a
Genomic Map