Assay Details
| Gene Symbol: Cdc25a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133571.1 | NP_598255.1 | 9 - 10 | 1168 | 70 | |
| XM_006243702.2 | XP_006243764.1 | 9 - 10 | 1390 | 70 | ||
| XM_006243703.1 | XP_006243765.1 | 10 - 11 | 941 | 70 | ||
| GenBank mRNA | D16236.1 | - | 9 - 10 | 1168 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdc25a
Genomic Map