Assay Details
| Gene Symbol: Cxcl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138522.1 | NP_612531.1 | 5 - 6 | 505 | 64 | |
| GenBank mRNA | D21095.1 | - | 5 - 6 | 505 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of leukocyte chemotaxis
chemotaxis
inflammatory response
immune response
G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
hemopoiesis
neutrophil chemotaxis
response to lipopolysaccharide
regulation of cell proliferation
chemokine-mediated signaling pathway
cellular response to lipopolysaccharide
chemokine activity
CXCR chemokine receptor binding
positive regulation of leukocyte chemotaxis
chemotaxis
inflammatory response
immune response
G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
hemopoiesis
neutrophil chemotaxis
response to lipopolysaccharide
regulation of cell proliferation
chemokine-mediated signaling pathway
cellular response to lipopolysaccharide
chemokine activity
CXCR chemokine receptor binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P052017 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P255029 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204683 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316372 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112506 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110822 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950579 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781117 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920478 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387406 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497146 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23635 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138845 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P052017 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P255029 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P255029 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P052017 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P255029 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P052017 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P255029 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P363116 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 36535 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10775913 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10775906 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10775907 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10775909 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10775908 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10775911 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10775912 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10775905 | - | - |
| Rat Genome 230 2.0 Array | 1388033_at | - | - |
| Rat Genome 230 2.0 Array | 1393182_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1388033_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1370633_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1370633_at | - | - |
| Rat Genome 230 2.0 Array | 1388032_PM_a_at | - | - |
| Rat Genome 230 2.0 Array | 1370634_PM_x_at | - | - |
| Rat Genome 230 2.0 Array | 1370634_x_at | - | - |
| Rat Genome 230 2.0 Array | 1393182_at | - | - |
| Rat Genome 230 2.0 Array | 1388032_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1388032_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1393182_at | - | - |
| Affymetrix HT Rat Focus Array | 1388033_at | - | - |
| Affymetrix HT Rat Focus Array | 1370633_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1388033_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1388032_PM_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1370633_at | - | - |
| Affymetrix HT Rat Focus Array | 1393182_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1370634_x_at | - | - |
| Affymetrix HT Rat Focus Array | 1370634_PM_x_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1400011064.rn.1 | TC1400001507.rn.1 | RTA TC |
| Rat Expression Set 230 | 1370633_at | - | - |
| Rat Expression Set 230 | 1370633_PM_at | - | - |
| Rat Expression Set 230 | 1370634_x_at | - | - |
| Rat Expression Set 230 | 1388032_a_at | - | - |
| Rat Expression Set 230 | 1370634_PM_x_at | - | - |
| Rat Expression Set 230 | 1388033_PM_at | - | - |
| Rat Expression Set 230 | 1388033_at | - | - |
| Rat Expression Set 230 | 1388032_PM_a_at | - | - |
| Rat Expression Set 230 | 1393182_PM_at | - | - |
| Rat Expression Set 230 | 1393182_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10775905 | - | - |
| Rat Genome U34 Set | D21095_at | - | - |
| Rat Genome U34 Set | rc_AA849026_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20731469 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1368177 | - | - |
| RatRef-12 Expression BeadChip | ILMN_58190 | - | - |

Cxcl3
Genomic Map