Assay Details
| Gene Symbol: Gpr149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138891.1 | NP_620246.1 | 3 - 4 | 2157 | 72 | |
| GenBank mRNA | AY030276.1 | - | 3 - 4 | 2157 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
preantral ovarian follicle growth
antral ovarian follicle growth
G-protein coupled receptor signaling pathway, coupled to cyclic nucleotide second messenger
neuropeptide signaling pathway
chemical synaptic transmission
negative regulation of ovulation
G-protein coupled receptor activity
neuropeptide binding
preantral ovarian follicle growth
antral ovarian follicle growth
G-protein coupled receptor signaling pathway, coupled to cyclic nucleotide second messenger
neuropeptide signaling pathway
chemical synaptic transmission
negative regulation of ovulation
G-protein coupled receptor activity
neuropeptide binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P205869 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434348 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972292 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905800 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937851 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952279 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99331 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413900 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505790 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21598 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411715 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281092 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149180 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701751 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595543 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724671 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701026 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942586 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911003 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564932 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318489 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320970 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816206 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103282 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603952 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924665 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2766 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939524 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595117 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781266 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38893 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101156 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85019 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121719 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63800 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9488 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P205869 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P205869 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P205869 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P205869 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 38234 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 23065 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 11164 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 20763 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 13504 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 69516 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10823413 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823415 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823416 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823414 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10823412 | - | - |
| Rat Genome 230 2.0 Array | 1370572_at | - | - |
| Rat Genome 230 2.0 Array | 1370572_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1370572_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1370572_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0200025290.rn.1 | TC0200003774.rn.1 | RTA TC |
| Rat Expression Set 230 | 1370572_at | - | - |
| Rat Expression Set 230 | 1370572_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823412 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21192535 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353576 | - | - |
| RatRef-12 Expression BeadChip | ILMN_70083 | - | - |

Gpr149
Genomic Map