Assay Details
| Gene Symbol: Dcps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153302.1 | NP_695214.1 | 4 - 5 | 628 | 64 | |
| GenBank mRNA | AF411249.1 | - | 4 - 5 | 628 | 64 | |
| BC089773.1 | - | 4 - 5 | 623 | 64 | ||
| BC105900.1 | - | 4 - 5 | 633 | 7377732 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P107097 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278026 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707607 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154652 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109433 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897353 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593807 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153138 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600067 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59328 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891948 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757172 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73661 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236635 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338397 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633523 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368713 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45092 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610695 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19636 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392939 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948248 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641584 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41180 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582765 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813708 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224519 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941779 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528750 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600852 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72929 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877831 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723601 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141865 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12437 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17683 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74613 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79570 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62793 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91337 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P107097 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P107097 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P107097 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P107097 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 57356 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 52904 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 85439 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 35034 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 4239 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10916033 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10916029 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10916030 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10916032 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10916034 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10916031 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10916028 | - | - |
| Rat Genome 230 2.0 Array | 1374583_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1390185_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1381527_at | - | - |
| Rat Genome 230 2.0 Array | 1390185_at | - | - |
| Rat Genome 230 2.0 Array | 1374583_at | - | - |
| Rat Genome 230 2.0 Array | 1381527_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1390185_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1390185_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0800030271.rn.1 | TC0800003676.rn.1 | RTA TC |
| Rat Expression Set 230 | 1390185_at | - | - |
| Rat Expression Set 230 | 1390185_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10916028 | - | - |
| Rat Genome U34 Set | rc_AI072775_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21007956 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1362522 | - | - |
| RatRef-12 Expression BeadChip | ILMN_61922 | - | - |

Dcps
Genomic Map