Assay Details
| Gene Symbol: Gnat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173139.1 | NP_775162.1 | 3 - 4 | 414 | 79 | |
| GenBank mRNA | X65747.1 | - | 3 - 4 | 414 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
detection of chemical stimulus involved in sensory perception of bitter taste
G-protein coupled receptor signaling pathway
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
visual perception
metabolic process
detection of visible light
response to nicotine
sensory perception of taste
sensory perception of bitter taste
sensory perception of sweet taste
sensory perception of umami taste
G-protein coupled receptor binding
GTPase activity
GTP binding
G-protein coupled photoreceptor activity
G-protein beta/gamma-subunit complex binding
metal ion binding
detection of chemical stimulus involved in sensory perception of bitter taste
G-protein coupled receptor signaling pathway
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
visual perception
metabolic process
detection of visible light
response to nicotine
sensory perception of taste
sensory perception of bitter taste
sensory perception of sweet taste
sensory perception of umami taste
G-protein coupled receptor binding
GTPase activity
GTP binding
G-protein coupled photoreceptor activity
G-protein beta/gamma-subunit complex binding
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P433147 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808977 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4280 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58859 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547899 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104075 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87570 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246510 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202270 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837772 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947538 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798821 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708193 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127994 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960776 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536875 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345705 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711017 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554669 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362913 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206198 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903381 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404251 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177923 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835149 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762436 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409578 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298379 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168267 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244876 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58818 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60408 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23104 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P433147 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P433147 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P433147 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P433147 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 11065 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 9902 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 89439 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 31195 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10860319 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10860318 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10860315 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10860320 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10860321 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10860317 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10860322 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10860324 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10860323 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10860316 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10860314 | - | - |
| Rat Genome 230 2.0 Array | 1388223_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1388223_at | - | - |
| Affymetrix HT Rat Focus Array | 1388223_at | - | - |
| Affymetrix HT Rat Focus Array | 1388223_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0400016038.rn.1 | TC0400002259.rn.1 | RTA TC |
| Rat Expression Set 230 | 1388223_PM_at | - | - |
| Rat Expression Set 230 | 1388223_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10860314 | - | - |
| Rat Genome U34 Set | X65747_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21090772 | - | - |

Gnat3
Genomic Map