Assay Details
| Gene Symbol: Cela2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012553.2 | NP_036685.1 | 3 - 4 | 249 | 65 | |
| GenBank mRNA | FQ226773.1 | - | 3 - 4 | 251 | 65 | |
| FQ227652.1 | - | 3 - 4 | 251 | 65 | ||
| FQ228000.1 | - | 3 - 4 | 251 | 65 | ||
| FQ228239.1 | - | 2 - 3 | 132 | 65 | ||
| V01233.1 | - | 3 - 4 | 249 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cela2a
Genomic Map