Assay Details
| Gene Symbol: Tpsb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019180.2 | NP_062053.2 | 2 - 3 | 245 | 63 | |
| GenBank mRNA | D38455.1 | - | 3 - 4 | 255 | 63 | |
| U67909.1 | - | 2 - 3 | 246 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tpsb2
Genomic Map