Assay Details
| Gene Symbol: Arf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024149.1 | NP_077063.1 | 1 - 2 | 162 | 99 | |
| GenBank mRNA | BC087692.1 | - | 1 - 2 | 124 | 7310424 | 99 |
| L12384.1 | - | 1 - 2 | 162 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arf5
Genomic Map