Assay Details
| Gene Symbol: Ap2m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053837.1 | NP_446289.1 | 1 - 2 | 132 | 65 | |
| GenBank mRNA | BC087724.1 | - | 139 | 7320615 | 65 | |
| M23674.1 | - | 1 - 2 | 132 | 65 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ap2m1
Genomic Map