Assay Details
| Gene Symbol: Tff2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053844.1 | NP_446296.1 | 1 - 2 | 107 | 93 | |
| GenBank mRNA | M97255.1 | - | 1 - 2 | 107 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tff2
Genomic Map