Assay Details
| Gene Symbol: Slc25a20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053965.2 | NP_446417.2 | 2 - 3 | 302 | 61 | |
| GenBank mRNA | BC081749.1 | - | 2 - 3 | 302 | 7134670 | 61 |
| FQ209681.1 | - | 2 - 3 | 258 | 61 | ||
| X97831.1 | - | 2 - 3 | 259 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc25a20
Genomic Map