Assay Details
| Gene Symbol: Impg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023958.1 | NP_076448.1 | 15 - 16 | 2373 | 60 | |
| GenBank mRNA | AB047843.1 | - | 15 - 16 | 2373 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Impg1
Genomic Map