Assay Details
| Gene Symbol: Tgfb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031131.1 | NP_112393.1 | 3 - 4 | 903 | 67 | |
| XM_006250448.2 | XP_006250510.1 | 2 - 3 | 1729 | 67 | ||
| GenBank mRNA | AF135598.1 | - | 2 - 3 | 513 | 67 | |
| AF153012.2 | - | 3 - 4 | 903 | 67 | ||
| AF153013.1 | - | 2 - 3 | 819 | 67 | ||
| AJ132718.1 | - | 2 - 3 | 527 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tgfb2
Genomic Map