Assay Details
| Gene Symbol: Cdkn2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031550.1 | NP_113738.1 | 1 - 2 | 167 | 73 | |
| GenBank mRNA | AF474976.1 | - | 1 - 2 | 143 | 73 | |
| AF474977.1 | - | 1 - 2 | 134 | 73 | ||
| AY679728.1 | - | 1 - 2 | 143 | 73 | ||
| L81167.1 | - | 1 - 2 | 167 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdkn2a
Genomic Map