Assay Details
| Gene Symbol: Sult1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031834.1 | NP_114022.1 | 6 - 7 | 652 | 97 | |
| GenBank mRNA | AF394783.1 | - | 5 - 6 | 575 | 97 | |
| FQ209630.1 | - | 6 - 7 | 661 | 97 | ||
| FQ210886.1 | - | 6 - 7 | 657 | 97 | ||
| FQ214244.1 | - | 6 - 7 | 657 | 97 | ||
| FQ218459.1 | - | 6 - 7 | 657 | 97 | ||
| FQ218506.1 | - | 6 - 7 | 666 | 98 | ||
| FQ218658.1 | - | 6 - 7 | 656 | 97 | ||
| FQ219178.1 | - | 6 - 7 | 657 | 97 | ||
| FQ219521.1 | - | 6 - 7 | 657 | 97 | ||
| FQ219542.1 | - | 6 - 7 | 657 | 97 | ||
| FQ219948.1 | - | 6 - 7 | 657 | 97 | ||
| L19998.1 | - | 6 - 7 | 652 | 97 | ||
| X52883.1 | - | 6 - 7 | 640 | 97 | ||
| X68640.1 | - | 5 - 6 | 557 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sult1a1
Genomic Map