Assay Details
| Gene Symbol: Rab11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032617.2 | NP_116006.1 | 3 - 4 | 480 | 87 | |
| GenBank mRNA | AF286534.1 | - | 3 - 4 | 486 | 87 | |
| BC062041.1 | - | 3 - 4 | 480 | 5598168 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rab11b
Genomic Map