Assay Details
| Gene Symbol: Gna15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053542.1 | NP_445994.1 | 1 - 2 | 306 | 115 | |
| XM_006241021.2 | XP_006241083.1 | 1 - 2 | 681 | 115 | ||
| GenBank mRNA | AB015308.1 | - | 1 - 2 | 381 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gna15
Genomic Map