Assay Details
| Gene Symbol: S100a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053681.1 | NP_446133.1 | 2 - 3 | 192 | 118 | |
| XM_006232558.2 | XP_006232620.1 | 2 - 3 | 395 | 118 | ||
| XM_006232559.2 | XP_006232621.1 | 2 - 3 | 1149 | 118 | ||
| XM_006232560.2 | XP_006232622.1 | 2 - 3 | 1598 | 118 | ||
| XM_006232563.2 | XP_006232625.1 | 2 - 3 | 180 | 118 | ||
| XM_008761122.1 | XP_008759344.1 | 3 - 4 | 627 | 118 | ||
| XM_008761123.1 | XP_008759345.1 | 2 - 3 | 670 | 118 | ||
| GenBank mRNA | AF140231.1 | - | 2 - 3 | 192 | 118 | |
More Information
Updated Mapping Information
Set Membership: |
|

S100a3
Genomic Map