Assay Details
| Gene Symbol: Casp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053736.2 | NP_446188.2 | 1 - 2 | 270 | 93 | |
| GenBank mRNA | AY029283.1 | - | 1 - 2 | 293 | 93 | |
| BC104720.1 | - | 1 - 2 | 270 | 7382967 | 93 | |
| FQ227402.1 | - | 1 - 2 | 307 | 93 | ||
| FQ232279.1 | - | 1 - 2 | 307 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Casp4
Genomic Map