Assay Details
| Gene Symbol: Plekhb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172033.2 | NP_742030.1 | 4 - 5 | 474 | 59 | |
| XM_006229787.2 | XP_006229849.1 | 5 - 6 | 712 | 59 | ||
| XM_006229788.2 | XP_006229850.1 | 4 - 5 | 873 | 59 | ||
| XM_006229789.2 | XP_006229851.1 | 5 - 6 | 783 | 59 | ||
| XM_006229791.2 | XP_006229853.1 | 4 - 5 | 1345 | 59 | ||
| XM_008759795.1 | XP_008758017.1 | 4 - 5 | 869 | 59 | ||
| GenBank mRNA | AF081582.1 | - | 4 - 5 | 474 | 59 | |
| AF118562.1 | - | 4 - 5 | 444 | 59 | ||
| BC078704.1 | - | 4 - 5 | 504 | 7124581 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plekhb1
Genomic Map