Assay Details
| Gene Symbol: Crybb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012937.1 | NP_037069.1 | 2 - 3 | 204 | 84 | |
| GenBank mRNA | X16072.1 | - | 2 - 3 | 204 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Crybb2
Genomic Map