Assay Details
| Gene Symbol: Il2rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013195.1 | NP_037327.1 | 9 - 10 | 1015 | 137 | |
| XM_006241970.2 | XP_006242032.1 | 9 - 10 | 1444 | 137 | ||
| GenBank mRNA | M55050.1 | - | 9 - 10 | 1015 | 137 | |
More Information
Updated Mapping Information
Set Membership: |
|

Il2rb
Genomic Map