Assay Details
| Gene Symbol: Glyatl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134330.2 | NP_599157.2 | 2 - 3 | 118 | 88 | |
| GenBank mRNA | AB019693.1 | - | 184 | 88 | ||
| BC078701.1 | - | 2 - 3 | 118 | 7130696 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Glyatl2
Genomic Map