Assay Details
| Gene Symbol: Spata19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134384.1 | NP_599211.1 | 3 - 4 | 259 | 75 | |
| GenBank mRNA | AB057408.1 | - | 3 - 4 | 259 | 75 | |
| BC081729.1 | - | 3 - 4 | 276 | 7132055 | 75 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Spata19
Genomic Map