Assay Details
| Gene Symbol: Spata19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134384.1 | NP_599211.1 | 5 - 6 | 423 | 96 | |
| GenBank mRNA | AB057408.1 | - | 5 - 6 | 423 | 96 | |
| BC081729.1 | - | 5 - 6 | 440 | 7132055 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spata19
Genomic Map