Assay Details
| Gene Symbol: Ifng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138880.2 | NP_620235.1 | 2 - 3 | 193 | 142 | |
| GenBank mRNA | AF010466.1 | - | 2 - 3 | 193 | 142 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ifng
Genomic Map