Assay Details
| Gene Symbol: Dhrs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153315.2 | NP_695227.2 | 1 - 2 | 260 | 98 | |
| GenBank mRNA | AB062758.1 | - | 1 - 2 | 76 | 98 | |
| BC070961.1 | - | 1 - 2 | 260 | 7099044 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dhrs4
Genomic Map