Assay Details
| Gene Symbol: Slc12a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153625.1 | NP_705889.1 | 3 - 4 | 436 | 65 | |
| XM_006248422.2 | XP_006248484.1 | 4 - 5 | 619 | 65 | ||
| XM_006248423.2 | XP_006248485.1 | 4 - 5 | 584 | 65 | ||
| XM_006248424.2 | XP_006248486.1 | 4 - 5 | 868 | 65 | ||
| XM_006248425.2 | XP_006248487.1 | 4 - 5 | 457 | 65 | ||
| XM_006248426.2 | XP_006248488.1 | 4 - 5 | 619 | 65 | ||
| XM_006248427.2 | XP_006248489.1 | 4 - 5 | 619 | 65 | ||
| XM_006248429.2 | XP_006248491.1 | 4 - 5 | 619 | 65 | ||
| XM_008768704.1 | XP_008766926.1 | 4 - 5 | 619 | 65 | ||
| XM_008768706.1 | XP_008766928.1 | 4 - 5 | 619 | 65 | ||
| GenBank mRNA | AB024430.1 | - | 3 - 4 | 436 | 65 | |
| AB024431.1 | - | 3 - 4 | 436 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc12a8
Genomic Map