Assay Details
| Gene Symbol: Ppif | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172243.1 | NP_758443.1 | 2 - 3 | 256 | 125 | |
| GenBank mRNA | BC086977.1 | - | 2 - 3 | 289 | 7193156 | 125 |
| U68544.1 | - | 2 - 3 | 256 | 125 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppif
Genomic Map