Assay Details
| Gene Symbol: Ppif | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172243.1 | NP_758443.1 | 3 - 4 | 340 | 67 | |
| GenBank mRNA | BC086977.1 | - | 3 - 4 | 373 | 7193156 | 67 |
| U68544.1 | - | 3 - 4 | 340 | 67 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ppif
Genomic Map