Assay Details
| Gene Symbol: Gclc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012815.2 | NP_036947.1 | 3 - 4 | 505 | 65 | |
| GenBank mRNA | BC081702.1 | - | 3 - 4 | 505 | 7110739 | 65 |
| J05181.1 | - | 3 - 4 | 477 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gclc
Genomic Map