Assay Details
| Gene Symbol: Sod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017051.2 | NP_058747.1 | 3 - 4 | 421 | 64 | |
| GenBank mRNA | BC070913.1 | - | 3 - 4 | 421 | 7104698 | 64 |
| Y00497.1 | - | 3 - 4 | 349 | 64 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sod2
Genomic Map