Assay Details
| Gene Symbol: Neu2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017130.2 | NP_058826.2 | 1 - 2 | 180 | 88 | |
| GenBank mRNA | D16300.1 | - | 1 - 2 | 180 | 88 | |
| FQ213221.1 | - | 1 - 2 | 143 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the D16300.1, NM_017130.2 transcripts.
Set Membership: |
|

Neu2
Genomic Map