Assay Details
| Gene Symbol: Clcn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017137.1 | NP_058833.1 | 533 | 63 | ||
| XM_006248562.2 | XP_006248624.1 | 1026 | 63 | |||
| XM_006248563.2 | XP_006248625.1 | 1026 | 63 | |||
| GenBank mRNA | X64139.1 | - | 533 | 63 | ||
| Gene Symbol: LOC100912455 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751081.3 | XP_003751129.1 | 3 - 4 | 1026 | 63 | |
| XM_006248554.2 | XP_006248616.1 | 3 - 4 | 1026 | 63 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Clcn2
Genomic Map